Testosterone ELISA Kit
Testosterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Testosterone in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples.
Testosterone ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of Testosterone in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. The 96-well microtiter plate has been pre-coated with Testosterone antigen. During the incubation, Testosterone present in the samples or standards competes with the fixed amount of immobilized Testosterone for binding sites on the Biotinylated Anti-Testosterone Antibody. The more Testosterone present in a sample or standard, the less Biotinylated Anti-Testosterone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Testosterone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Testosterone present in each sample or standard. The concentration of Testosterone can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
: This is a Ready-To-Use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Description:Testosterone ELISA Kit
- Primary antibody reactivity:Other
- Sample type:Serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples
- Sample volume:50 μl
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Competitive
- Time to Results:2 h
- Cross reactivity:Testosterone ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Testosterone. No significant cross-reactivity or interference between Testosterone and analogues was observed.
- Cat. No.:ANTIA326905-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:18,75 pg/ml
- Format:Pre-coated
- Target protein:Testosterone
- Detection limit:Intra-assay CV: <8%, Inter-assay CV: <10%
- Regulatory status:RUO



