Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse eNOS ELISA Kit

Mouse eNOS ELISA Kit

New Product
Supplier:   Antibodies.com
Mouse eNOS ELISA Kit
Supplier:  Antibodies.com
New Product
Certificates

2 Options Available Below

Product Details & Documents

Mouse eNOS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse eNOS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids.

Mouse eNOS ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse eNOS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. An antibody specific for eNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the eNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-eNOS Antibody, which binds the captured eNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of eNOS captured in each well. The concentration of eNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ANTIA1813-48
  • ANTIA1813-96
  • Target protein:
  • eNOS
  • eNOS
  • Primary antibody reactivity:
  • Mouse
  • Mouse
  • Assay Type:
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • Multiple steps
  • Multiple steps
  • Sensitivity:
  • 0,67 ng/ml
  • 0,67 ng/ml
  • Detection range:
  • 1,56 - 100 ng/ml
  • 1,56 - 100 ng/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Detection limit:
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Sample type:
  • Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
  • Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
  • Time to Results:
  • 4 h 30 min
  • 4 h 30 min
  • Regulatory status:
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C
  • Shelf life:
  • Store for 6 months at 4 °C
  • Store for 6 months at 4 °C

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ANTIA1813-48
    Mouse eNOS ELISA Kit
    Mouse eNOS ELISA Kit
    Log in for availability
    Log in for availability
    1 * 48 Tests
    1 * 48 Tests - 565,00 €
    565,00 €
    1 * 48 Tests
       565,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA1813-48
    Target protein
    eNOS
    Primary antibody reactivity
    Mouse
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,67 ng/ml
    Detection range
    1,56 - 100 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
    Time to Results
    4 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product
  • ANTIA1813-96
    Mouse eNOS ELISA Kit
    Mouse eNOS ELISA Kit
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 760,00 €
    760,00 €
    1 * 96 Tests
       760,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA1813-96
    Target protein
    eNOS
    Primary antibody reactivity
    Mouse
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,67 ng/ml
    Detection range
    1,56 - 100 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
    Time to Results
    4 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product