Mouse Glutaredoxin 5 ELISA Kit
Mouse Glutaredoxin 5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Glutaredoxin 5 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples.
Mouse Glutaredoxin 5 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Glutaredoxin 5 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. An antibody specific for Glutaredoxin 5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Glutaredoxin 5 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Glutaredoxin 5 Antibody, which binds the captured Glutaredoxin 5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Glutaredoxin 5 captured in each well. The concentration of Glutaredoxin 5 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
: This is a Ready-To-Use ELISA kit that contains: Pre-Coated 96 Well Microplate, Lyophilized Standard, Sample Dilution Buffer, Biotinylated Detection Antibody (100X), Antibody Dilution Buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC Dilution Buffer, TMB Substrate, Stop Solution, Wash Buffer (25X), Plate Sealers, Foil Pouch, and Protocol Manual.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Description:Mouse Glutaredoxin 5 ELISA kit
- Sample type:Serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples
- Sample volume:100 μl
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 hours
- Cross reactivity:Mouse Glutaredoxin 5 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Glutaredoxin 5. No significant cross-reactivity or interference between Glutaredoxin 5 and analogues was observed.
- Cat. No.:ANTIA326590-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:9,375 pg/ml
- Format:Pre-coated
- Target protein:Glutaredoxin 5
- Detection limit:Intra-assay CV: <8%, Inter-assay CV: <10%
- Regulatory status:RUO



