Human Amyloid beta 1-40 ELISA Kit
Catalog # ABCAAB193692-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Amyloid beta 1-40
- Description:Human Amyloid beta 1-40 ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:0,1 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,1 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human Amyloid beta 1-40 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Amyloid beta 1-40 in Human in plasma, cell culture supernatant, Serum samples.