Order Entry
Export
ContactUsLinkComponent
Human IL-13 ELISpot Kit
Human IL-13 ELISpot Kit
Supplier:  Abcam
Certificates
undefined
Human IL-13 ELISpot Kit
Supplier:  Abcam

Specifications

  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Antigen symbol:
    IL-13
  • Product type:
    Pre-coated

Specifications