Human BCAM (CD239) ELISA Kit
Catalog # ABCAAB304935-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human BCAM (CD239) ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human BCAM (CD239) ELISA kit is a single-wash 90-min Simplestep used to quantify Human BCAM (CD239) with a sensitivity of 5,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.