Pig Lipocalin-2 ELISA Kit
Catalog # ABCAAB207924-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Pig Lipocalin-2 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Pig Lipocalin-2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig Lipocalin-2 in Pig in urine, plasma, serum samples.