Order Entry
Export
ContactUsLinkComponent
Human Glucose-6-Phosphate Isomerase ELISA Kit
Human Glucose-6-Phosphate Isomerase ELISA Kit
Catalog # ABCAAB171575-96
Supplier:  Abcam
CAS Number:  
undefined
Human Glucose-6-Phosphate Isomerase ELISA Kit
Catalog # ABCAAB171575-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Glucose 6 Phosphate Isomerase
  • Description:
    Human Glucose-6-Phosphate Isomerase ELISA kit
  • Sample type:
    Tissue homogenate, serum, cell lysate
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    61 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human Glucose-6-Phosphate Isomerase ELISA kit is a single-wash 90-min Simplestep used to quantify Human Glucose-6-Phosphate Isomerase with a sensitivity of 61 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Cited in over 5 citations.