Human SHP2 ELISA Kit
Catalog # ABCAAB314344-96TST
Supplier: Abcam
CAS Number:
Specifications
- Description:Human SHP2 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human SHP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human SHP2 in Human in Cell culture supernatant, Plasma, Serum samples.