Order Entry
Export
ContactUsLinkComponent
Human Fas Ligand ELISPOT Kit
Human Fas Ligand ELISPOT Kit
Supplier:  Abcam
Certificates
undefined
Human Fas Ligand ELISPOT Kit
Supplier:  Abcam

Specifications

  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Antigen name:
    Fas Ligand
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO

Specifications