Mouse IL-10 ELISA kit
Catalog # ABCAAB108870-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IL-10
- Description:Mouse IL-10 ELISA kit
- Sample type:Plasma, cell culture supernatant, serum, cell lysate, tissue
- Detection method:Colorimetric
- Time to Results:5 h 0 min
- Detection range:0,125 - 1 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,014 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-10 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-10 in Mouse in plasma, cell culture supernatant, Serum, Cell Lysate, Tissue samples.