Human IL-36 gamma ELISA (IL-1F9)
Catalog # ABCAAB267633-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-36 gamma ELISA (IL-1F9)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human IL-36 gamma ELISA (IL-1F9) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-36 gamma ELISA (IL-1F9) in Human in plasma, cell culture supernatant, serum samples.