Infliximab ELISA Kit
Catalog # ABCAAB237647-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Infliximab ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Infliximab ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Infliximab in Human in Plasma, Serum samples.