Swine Haptoglobin ELISA Kit
Catalog # ABCAAB223136-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Swine Haptoglobin ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Swine Haptoglobin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Swine Haptoglobin in Pig in citrate plasma, serum, urine samples.