Order Entry
Export
Orders LinkContactUsLinkComponent
 

96-Well Microplate Pack

96-Well Microplate pack, 10 pack
Catalog # ABCAAB210903-10
Supplier:   Abcam
96-Well Microplate Pack
Catalog # ABCAAB210903-10
Supplier:  Abcam
96-Well Microplate Pack
96-Well Microplate Pack

96-Well Microplate pack, 10 pack

Price
164,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

96-Well Microplate Pack (ab210903) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

 : RUO

Specifications
  • Packed:
    10 pack
  • Description:
    96-Well Microplate pack
  • Cat. No.:
    ABCAAB210903-10
  • Detection method:
    Multiple steps
  • Storage temperature:
    4 ℃
  • Well profile:
    Uncoated