Human TNF alpha ELISPOT Kit
Supplier: Abcam
Certificates
About this item
Human TNF alpha ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human TNF alpha production in a single cell suspension.
Specifications
- Species reactivity:Human
- Sample type:Suspension cells
- Form:Pre-coated
- Storage temperature:4 °C
- Processing time:Multiple steps
- Antigen symbol:TNF alpha
- Regulatory status:RUO