Human SOX2 ELISA Kit
ABCAAB245707-1X96
: Abcam
:
- Description:Human SOX2 ELISA kit
- Tests per kit:96 tests
Human SOX2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human SOX2 with a sensitivity of 31 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.