Human IL-5 ELISpot Pair
Catalog # ABCAAB281990-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-5 ELISpot pair
- Species reactivity:Human
- Sample type:Suspension cells
- Form:Uncoated
- Storage temperature:+4 ℃
- Processing time:Multiple steps
- Antigen symbol:IL-5
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human IL-5 ELISpot Pair is a ELISpot Pair for the sensitive detection of Human IL-5 production in a single cell suspension.