Mouse IL-16 ELISA Kit
Catalog # ABCAAB201282-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IL-16 ELISA kit
Specifications
About this item
Mouse IL-16 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse IL-16 with a sensitivity of 3.6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.