Human SMAC ELISA Kit
Catalog # ABCAAB155439-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human SMAC ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human SMAC ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human SMAC in Human in plasma, cell culture supernatant, serum samples.