Chicken IL-10 ELISA Kit
Catalog # ABCAAB273259-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Chicken IL-10 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Chicken IL-10 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Chicken IL-10 in Chicken in Cell culture supernatant, Serum, EDTA Plasma samples.