Rat IL-10 ELISA Kit
Catalog # ABCAAB108872-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat IL-10 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat IL-10 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat IL-10 in Rat in Cell culture supernatant samples.