Monkey IL-18 ELISA Kit
ABCAAB316257-96TST
: Abcam
New Product
:
- Description:Monkey IL-18 ELISA Kit
- Tests per kit:96 tests
Monkey IL-18 ELISA Kit is a single-wash 90-min Simplestep used to quantify Monkey IL-18 with a sensitivity of 3,069 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.