Order Entry
Export
Orders LinkContactUsLinkComponent
Human Hemoglobin ELISA Kit

Human Hemoglobin ELISA Kit

Human Hemoglobin ELISA kit, 96 tests
Catalog # ABCAAB157707-96
Supplier:   Abcam
Human Hemoglobin ELISA Kit
Catalog # ABCAAB157707-96
Supplier:  Abcam
CAS Number:  
Human Hemoglobin ELISA Kit
Human Hemoglobin ELISA Kit

Human Hemoglobin ELISA kit, 96 tests

CAS Number:  
Price
790,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Hemoglobin ELISA kit is a Sandwich ELISA used to quantify Human Hemoglobin with a sensitivity of 0,8445 ng/ml. Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader. Wide dynamic range - 6,25 to 200 ng/ml.

Specifications

Specifications

  • Description:
    Human Hemoglobin ELISA kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB157707-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,8445 ng/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Sample type:
    Plasma, Serum
  • Target protein:
    Hemoglobin
  • Detection range:
    6,25 - 200 ng/ml
  • Storage temperature:
    4 ℃
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications