Streptomycin ELISA Kit
Catalog # ABCAAB285226-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Streptomycin ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Streptomycin ELISA kit is a Competitive ELISA kit for the measurement of Streptomycin in tissue, milk samples.