Order Entry
Export
ContactUsLinkComponent
Neomycin ELISA Kit
Neomycin ELISA Kit
Catalog # ABCAAB285276-96
Supplier:  Abcam
CAS Number:  
undefined
Neomycin ELISA Kit
Catalog # ABCAAB285276-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Description:
    Neomycin ELISA kit
  • Tests per kit:
    100 assays

Specifications

About this item

A competitive ELISA kit for quantitate of neomycin in tissue, honey, milk, milk powder, liver.

  • Easy, convenient and time-saving method
  • Detection Range: 0,1 to 8,1 ppb
  • Sensitivity: <0,1 ppb
  • Detection limitation: 4 ppb

Neomycin is an aminoglycoside antibiotic found in many topical medications such as creams, ointments, and eyedrops. It is typically used as a topical preparation, such as neosporin. It can also be given orally, where it is usually combined with other antibiotics.

Neomycin, like other aminoglycosides, has been shown to be ototoxic causing tinnitus, hearing loss, and vestibular problems in a small number of patients. Standard techniques/instruments (HPLC or GC-MS) are utilised to detect neomycin.

However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of neomycin in various samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.