Mouse IL-28 ELISA Kit
Catalog # ABCAAB100708-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IL-28
- Description:Mouse IL-28 ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:2,46 - 600 pg/ml
- Storage temperature:4 °C
- Sensitivity:2 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-28 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-28 in Mouse in plasma, cell culture supernatant, Serum samples.