Canine IL-10 ELISA Kit
Catalog # ABCAAB193685-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Canine IL-10 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Canine IL-10 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Canine IL-10 in Dog in Plasma, cell culture supernatant, serum samples.
Certifications: RUO