Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human CD33 ELISA Kit

Human CD33 ELISA kit, 96 tests
Catalog # ABCAAB283542-1X96
Supplier:   Abcam
Human CD33 ELISA Kit
Catalog # ABCAAB283542-1X96
Supplier:  Abcam
CAS Number:  
Human CD33 ELISA Kit
Human CD33 ELISA Kit

Human CD33 ELISA kit, 96 tests

CAS Number:  
Price
785,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human CD33 ELISA kit is a single-wash 90-min Simplestep used to quantify Human CD33 with a sensitivity of 11.39 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human CD33 ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Heparin plasma, serum, cell lysate, EDTA plasma
  • Detection range:
    70,31 - 4500 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB283542-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    11,39 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    CD33
  • Regulatory status:
    RUO

Specifications