Human IL-32 ELISA Kit
Catalog # ABCAAB287166-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-32 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human IL-32 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IL-32 with a sensitivity of 5,84 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.