Mouse IL-33 ELISA Kit
Catalog # ABCAAB171181-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IL-33
- Description:Mouse IL-33 ELISA kit
- Sample type:Cell lysate, cell culture supernatant, edta plasma, heparin plasma, serum
- Detection method:Colorimetric
- Detection range:31,2 - 2000 pg/ml
- Storage temperature:4 °C
- Sensitivity:10 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-33 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-33 in Mouse in Cell Lysate, cell culture supernatant, EDTA plasma, heparin plasma, serum samples.