Order Entry
Export
ContactUsLinkComponent
Human IL-2 ELISPOT Kit
Human IL-2 ELISPOT Kit
Supplier:  Abcam
Certificates
undefined
Human IL-2 ELISPOT Kit
Supplier:  Abcam

Specifications

  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps, 3 hr 0 min
  • Antigen symbol:
    IL-2
  • Product type:
    Pre-coated

Specifications