Order Entry
Export
Orders LinkContactUsLinkComponent
Aldosterone ELISA kit

Aldosterone ELISA kit

Aldosterone ELISA kit, 96 tests
Catalog # ABCAAB136933-96
Supplier:   Abcam
Aldosterone ELISA kit
Catalog # ABCAAB136933-96
Supplier:  Abcam
CAS Number:  
Aldosterone ELISA kit
Aldosterone ELISA kit

Aldosterone ELISA kit, 96 tests

CAS Number:  
Price
805,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Aldosterone ELISA kit ab136933 is used to quantify Aldosterone with a sensitivity of 4.7 pg/ml. Competitive ELISA kit.

Specifications

Specifications

  • Description:
    Aldosterone ELISA kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB136933-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,7 pg/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human|Mammals|Mouse|Rat
  • Sample type:
    Urine, plasma, serum
  • Target protein:
    Aldosterone
  • Detection range:
    3,9 - 250 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Competitive

Specifications