Human IL-17 ELISA Kit
Catalog # ABCAAB119535-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-17 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human IL-17 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-17 in Human in Plasma, Cell culture extracts, Serum samples.