Mouse IgA ELISA Kit
Mouse IgA ELISA kit, 96 tests
Catalog # ABCAAB314603-96TST
New Product
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse IgA ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse IgA with a sensitivity of 2,2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
Specifications
Specifications
- Description:Mouse IgA ELISA kit
- Tests per kit:96 tests
- Primary antibody reactivity:Mouse
- Sample type:Serum, citrate plasma, EDTA plasma, heparin plasma
- Detection range:40,625 - 2600 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB314603-96TST
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:2,2 pg/ml
- Format:Pre-coated
- Target protein:IgA
- Regulatory status:RUO



