Complex I Human Protein Quantity Dipstick Assay Kit
Catalog # ABCAAB109722-48
Supplier: Abcam
CAS Number:
Specifications
- Description:Complex I Human Protein Quantity Dipstick Assay kit
- Method:Sandwich
- Species reactivity:Other;Human
- Sample type:Cell culture extracts, Tissue
- Antigen name:Complex I
- Storage temperature:4 °C
- Processing time:Multiple steps
- Product type:Pre-coated
- Tests per kit:48 tests
Specifications
About this item
Complex I Human Protein Quantity Dipstick Assay Kit is an immunoassay kit for the detection of Complex I Human Protein Quantity in Cow, Human in Cell culture extracts, Tissue samples.