Human FGFR3 ELISA Kit
ABCAAB214027-96
: Abcam
:
- Description:Human FGFR3 ELISA Kit
- Tests per kit:96 tests
Human FGFR3 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human FGFR3 with a sensitivity of 61 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.