Human IL-10 ELISA Kit High Sensitivity
Catalog # ABCAAB46059-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-10 ELISA kit High Sensitivity
- Tests per kit:96 tests
Specifications
About this item
Human IL-10 ELISA Kit High Sensitivity is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-10 High Sensitivity in Human in Plasma, Cell culture supernatant, Serum samples.