Human IP-10 ELISA Kit, Fluorescent
Catalog # ABCAAB229410-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IP-10 ELISA kit, Fluorescent
- Tests per kit:96 tests
Specifications
About this item
Human IP-10 ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 1,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader.