Order Entry
Export
Orders LinkContactUsLinkComponent
Human Factor IX ELISA Kit

Human Factor IX ELISA Kit

Human Factor IX ELISA kit, 96 tests
Catalog # ABCAAB108831-96
Supplier:   Abcam
Human Factor IX ELISA Kit
Catalog # ABCAAB108831-96
Supplier:  Abcam
CAS Number:  
Human Factor IX ELISA Kit
Human Factor IX ELISA Kit

Human Factor IX ELISA kit, 96 tests

CAS Number:  
Price
805,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Factor IX ELISA kit ab108831 is used to quantify Human Factor IX with a sensitivity of 20,6 pg/ml. Sandwich ELISA kit.

Specifications

Specifications

  • Description:
    Human Factor IX ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Plasma, Cell culture supernatant, Cerebral Spinal Fluid, Serum
  • Detection range:
    1,56 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 0 min
  • Cat. No.:
    ABCAAB108831-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,4 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Factor IX/PTC
  • Regulatory status:
    RUO

Specifications