Human ACY-1 ELISA
Catalog # ABCAAB277463-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human ACY-1 ELISA
Specifications
About this item
Human ACY-1 ELISA is a Sandwich (quantitative) ELISA kit for the measurement of Human ACY-1 ELISA in Human in cell culture supernatant, serum, EDTA plasma samples.