Human IL-32 alpha ELISA Kit
Catalog # ABCAAB267616-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-32 alpha ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human IL-32 alpha ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-32 alpha in Human in Plasma, Cell culture media, Serum samples.