Human CD80 ELISA Kit (B7-1)
Catalog # ABCAAB256392-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human CD80 ELISA kit (B7-1)
- Tests per kit:96 tests
Specifications
About this item
Human CD80 ELISA kit (B7-1) is a single-wash 90-min Simplestep used to quantify Human CD80 (B7-1) with a sensitivity of 31 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.