8 isoprostane ELISA Kit
Catalog # ABCAAB175819-96
Supplier: Abcam
CAS Number:
Specifications
- Description:8 isoprostane ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
8 isoprostane ELISA kit is a Competitive ELISA kit for the measurement of 8 isoprostane in urine, plasma, cell culture extracts, tissue, serum samples.