Order Entry
Export
ContactUsLinkComponent
Mouse Pro-Collagen I alpha 1 Matched Antibody Pair Kit
Mouse Pro-Collagen I alpha 1 Matched Antibody Pair Kit
Catalog # ABCAAB216791-5X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Pro-Collagen I alpha 1 Matched Antibody Pair Kit
Catalog # ABCAAB216791-5X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse Pro-Collagen I alpha 1 matched antibody pair kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    5×96 tests

Specifications

About this item

Mouse Pro-Collagen I alpha 1 Matched Antibody Pair kit is a kit for ELISA development for the measurement of Mouse Pro-Collagen I alpha 1 by sandwich ELISA method.