Human CD300e ELISA Kit
Catalog # ABCAAB313525-96TST
Supplier: Abcam
CAS Number:
Specifications
- Description:Human CD300e ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human CD300e ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human CD300e in Human in cell culture supernatant, plasma, serum samples.