Canine IgG ELISA Kit
Catalog # ABCAAB157701-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IgG
- Description:Canine IgG ELISA kit
- Sample type:Plasma, serum, other biological fluids
- Detection method:Colorimetric
- Detection range:25 - 800 ng/ml
- Storage temperature:4 °C
- Sensitivity:7,148 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Canine IgG ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine IgG in Dog in plasma, Serum, Other biological fluids samples.