Order Entry
Export
ContactUsLinkComponent
Human IL-18 R beta ELISA Kit
Human IL-18 R beta ELISA Kit
Catalog # ABCAAB113325-96
Supplier:  Abcam
CAS Number:  
undefined
Human IL-18 R beta ELISA Kit
Catalog # ABCAAB113325-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    IL-18R Beta/IL-18RAP
  • Description:
    Human IL-18 R beta ELISA kit
  • Sample type:
    Plasma, cell culture supernatant, serum
  • Detection method:
    Colorimetric
  • Detection range:
    12,3 - 3000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    12 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human IL-18 R beta ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-18 R beta in Human in plasma, cell culture supernatant, Serum samples.