Human Cadherin 13 ELISA
Catalog # ABCAAB309455-96TST
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Human Cadherin 13 ELISA
- Tests per kit:96 tests
Specifications
About this item
Human Cadherin 13 ELISA is a Sandwich (quantitative) ELISA kit for the measurement of Human Cadherin 13 ELISA in Human in Serum, Plasma, Cell culture supernatant samples.