Phospho-Phospholipase C gamma 1/PLC-gamma-1 (Y771) ELISA Kit
Catalog # ABCAAB279892-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:Phospholipase C gamma 1/PLC-gamma-1
- Description:Phospho-Phospholipase C gamma 1/PLC-gamma-1 (Y771) ELISA kit
- Sample type:Cell lysate
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Phospho-Phospholipase C gamma 1/PLC-gamma-1 (Y771) ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-Phospholipase C gamma 1/PLC-gamma-1 (Y771) in Human in Cell Lysate samples.